Specific Gravity Hydrometers
Proveedor: Amarell
H826508 H826512 H826506 H826640/DKD3 H826640/DKD1 H826640 H826786DKD3 H826788DKD3 H826790DKD3 H826502 H826504 H826510 H826514 H826516 H826628 H826630 H826632 H826634 H826636 H826638 H826642 H826644 H826646 H826648 H826760-FL H826764-FL H826780 H826782 H826784 H826786 H826788 H826790 H826792 H826910 H826912 H826914 H826916 H826918 H826920 H826922 H826924 H826926 H826928 H826930 H826932 H826934 H826936 H826938 H826942 H826940 H826500 H826786WC H826788WC
AMARH826508EA 51.2 EUR
AMARH826508 AMARH826512 AMARH826506 AMARH826640/DKD3 AMARH826640/DKD1 AMARH826640 AMARH826786DKD3 AMARH826788DKD3 AMARH826790DKD3 AMARH826502 AMARH826504 AMARH826510 AMARH826514 AMARH826516 AMARH826628 AMARH826630 AMARH826632 AMARH826634 AMARH826636 AMARH826638 AMARH826642 AMARH826644 AMARH826646 AMARH826648 AMARH826760-FL AMARH826764-FL AMARH826780 AMARH826782 AMARH826784 AMARH826786 AMARH826788 AMARH826790 AMARH826792 AMARH826910 AMARH826912 AMARH826914 AMARH826916 AMARH826918 AMARH826920 AMARH826922 AMARH826924 AMARH826926 AMARH826928 AMARH826930 AMARH826932 AMARH826934 AMARH826936 AMARH826938 AMARH826942 AMARH826940 AMARH826500 AMARH826786WC AMARH826788WC
Specific Gravity Hydrometers
Hidrómetros
These general-purpose hydrometers are available with a thermometer inside the body or without a thermometer.
  • Capillary prismatic
  • Reliable and simple
  • Paper scale with type designation
  • Shot weighted body
Información de suministro: Supplied in a transparent plastic case.