Precision thermometers
Proveedor: Amarell
L25994-FL/CC2 L25948-FL/CC3 L25992-FL/CC3 L25994-FL/CC3 L25902-FL/DKD3 L25902-FL/VC L25906-FL/DKD5 L25904-FL/DKD5 L25900-FL/WC L25900-FL L25900-FL/VC L25902-FL/WC L25902-FL-PTFE L25904-FL L25904-FL/VC L25904-FL/WC L25904-FL-PTFE L25906-FL L25906-FL/VC L25906-FL-PTFE L25908-FL L25908-FL/WC L25910-GA L25910-GA/VC L25910-GA/WC L25912-GA L25922-FL L25922-FL/VC L25922-FL/WC L25924-FL L25924-FL/WC L25924-FL-PTFE L25926-CB L25926-FL L25928-CB L25928-FL L25930-FL L25934-GA L25944-CB L25944-CB/WC L25944-FL L25944-FL/WC L25948-CB L25902-FL L25948-CB/WC L25948-FL L25948-FL/WC L25948-FL-PTFE L25950-CB L25952-CB L25952-CB/WC L25952-FL L25952-FL/WC L25954-FL/WC L25990-CB L25992-CB L25992-CB/WC L25992-FL L25992-FL/WC L25992-FL-PTFE L25994-CB L25994-CB/WC L25994-FL L25994-FL/WC L25994-FL-PTFE L25996-CB L25998-CB L26000-CB L26004-CB L26004-CB/WC L26004-FL L26006-CB L26012-CB L25992-FL/DKD3 L25992-FL/DKD4 L26004-CB/DKD4 L25992-CB/DKD4 L25948-FL/DKD5 L25952-FL/DKD6 L25902FLDKD3CL L25990-CB/WC L25994-FL/DKD3 L25908-FL/DKD3 L25930-FL/DKD4 L26006-FL/DKD3 L25904-FL/DKD1 L25992-GA/VC
AMARL25994-FL/CC2EA 295 EUR
AMARL25994-FL/CC2 AMARL25948-FL/CC3 AMARL25992-FL/CC3 AMARL25994-FL/CC3 AMARL25902-FL/DKD3 AMARL25902-FL/VC AMARL25906-FL/DKD5 AMARL25904-FL/DKD5 AMARL25900-FL/WC AMARL25900-FL AMARL25900-FL/VC AMARL25902-FL/WC AMARL25902-FL-PTFE AMARL25904-FL AMARL25904-FL/VC AMARL25904-FL/WC AMARL25904-FL-PTFE AMARL25906-FL AMARL25906-FL/VC AMARL25906-FL-PTFE AMARL25908-FL AMARL25908-FL/WC AMARL25910-GA AMARL25910-GA/VC AMARL25910-GA/WC AMARL25912-GA AMARL25922-FL AMARL25922-FL/VC AMARL25922-FL/WC AMARL25924-FL AMARL25924-FL/WC AMARL25924-FL-PTFE AMARL25926-CB AMARL25926-FL AMARL25928-CB AMARL25928-FL AMARL25930-FL AMARL25934-GA AMARL25944-CB AMARL25944-CB/WC AMARL25944-FL AMARL25944-FL/WC AMARL25948-CB AMARL25902-FL AMARL25948-CB/WC AMARL25948-FL AMARL25948-FL/WC AMARL25948-FL-PTFE AMARL25950-CB AMARL25952-CB AMARL25952-CB/WC AMARL25952-FL AMARL25952-FL/WC AMARL25954-FL/WC AMARL25990-CB AMARL25992-CB AMARL25992-CB/WC AMARL25992-FL AMARL25992-FL/WC AMARL25992-FL-PTFE AMARL25994-CB AMARL25994-CB/WC AMARL25994-FL AMARL25994-FL/WC AMARL25994-FL-PTFE AMARL25996-CB AMARL25998-CB AMARL26000-CB AMARL26004-CB AMARL26004-CB/WC AMARL26004-FL AMARL26006-CB AMARL26012-CB AMARL25992-FL/DKD3 AMARL25992-FL/DKD4 AMARL26004-CB/DKD4 AMARL25992-CB/DKD4 AMARL25948-FL/DKD5 AMARL25952-FL/DKD6 AMARL25902FLDKD3CL AMARL25990-CB/WC AMARL25994-FL/DKD3 AMARL25908-FL/DKD3 AMARL25930-FL/DKD4 AMARL26006-FL/DKD3 AMARL25904-FL/DKD1 AMARL25992-GA/VC
Precision thermometers
Termómetros Analogue Thermometers
These total immersion thermometers, with enclosed scale, are suitable for government testing and official calibration.
  • Milk glass scale
  • Fine subdivision up to 0,1 °C
Available with quality certificates.
Envase: Packaged in transparent square plastic case.