Precision thermometer
Proveedor: Amarell
M235000-FL/VC M235000-FL M235002-FL M235004-GA
AMARM235000-FL/VCEA 293 EUR
AMARM235000-FL/VC AMARM235000-FL AMARM235002-FL AMARM235004-GA
Precision thermometer
Termómetros Analogue Thermometers
Dropping point thermometers according to Ubbelohde.
  • Richter-top finish
  • Colourless, prismatic capillary
  • Red or gallium filled
These enclosed milk glass scale thermometers are suitable for government verification or for government calibration.
Certificaciones: Conforms to DIN 12785.
Información para pedidos: Available with or without verification certificate.
Envase: Packed in a transparent square plastic case.