Monos industriales, AlphaTec® 2300 COMFORT, modelo 129
Proveedor: Ansell
WY23B-00129-03 WY23B-00129-04 WY23B-00129-05 WY23B-00129-06 WY23B-00129-07
ANSEWY23B-00129-03EA 380 EUR
ANSEWY23B-00129-03 ANSEWY23B-00129-04 ANSEWY23B-00129-05 ANSEWY23B-00129-06 ANSEWY23B-00129-07
Monos industriales, AlphaTec® 2300 COMFORT, modelo 129
Prendas desechables
These overalls have been engineered for workers involved in composites manufacturing and related industries.
  • Three-piece hood - ensures a good fit with half and full face respirators
  • Elasticated hood, wrists, waist and ankles
  • Finger loops, prevent sleeve movement whilst working above your head
  • Two-way front zipper with resealable storm flap
  • Breathable SMS back panel
  • Silicone-free: Critical in spray painting applications
  • Fabric construction ensures that there is no risk of delamination should a wearer make contact with a tacky surface
The criticalMore Product Information
Order Now
ESPECIFICACIONES
Modelo 129
Environmentally Preferable
Tamaño de cierre Con cremallera
Antiestático +
Género
Seam type Bound
Aplicaciones de vestimenta Chemical protection,General Purpose,Industrial
Peso 62 g/m²