Human DNAJB8 ELISA Kit

Proveedor: ANTIBODIES.COM

A246932-96
ANTIA246932-96EA 635 EUR
ANTIA246932-96
Human DNAJB8 ELISA Kit
Ensayos ELISAs

Human DNAJB8 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human DNAJB8 in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready-to-use ELISA kit
  • Detection range: 0,313 to 20 ng/ml
  • Sensitivity: 0,188 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra - assay: CV <8%, Inter - assay: CV <10%


Human DNAJB8 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human DNAJB8 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for DNAJB8 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the DNAJB8 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-DNAJB8 antibody, which binds the captured DNAJB8 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of DNAJB8 captured in each well. The concentration of DNAJB8 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Información para pedidos: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

New Product

Order Now

ESPECIFICACIONES

Environmentally Preferable
Objetivo DNAJB8
Primary antibody reactivity Human
Tipo de ensayo Sandwich
Principio de ensayo Quantitative
Huésped
Formato Pre-coated
Duración del ensayo Multiple steps
Sensibilidad 0,188 ng/ml
Rango de detección 0,313 - 20 ng/ml
Método de detección Colorimetric
Reactividad cruzada Human DNAJB8 ELISA Kit exhibits high specificity and excellent specificity for the detection of human DNAJB8. No significant cross-reactivity or interference between DNAJB8 and analogues was observed.
Tipo de la muestra Serum, plasma, tissue homogenates, and other biological fluids
Volumen de la muestra 100 μl
Tiempo para resultados 4 h 30 min
Regulatory status RUO
Temperatura de almacenamiento 4 °C
Shelf life Store for 6 months at 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR