Anti-p23 Mouse Monoclonal Antibody [Clone: JJ6]

Proveedor: ANTIBODIES.COM
A304948-100
ANTIA304948-100EA 405 EUR
ANTIA304948-100
Anti-p23 Mouse Monoclonal Antibody [Clone: JJ6]
Anticuerpos
Mouse monoclonal [JJ6] antibody to p23 for WB, IHC, ICC/IF, IP, ELISA and antibody microarray with samples derived from human, mouse, rabbit, chicken, guinea pig and yeast.

  • Validated applications: WB, IHC, ICC/IF, IP, ELISA, Antibody microarray
  • Recommended dilutions: WB: 1:2000 IHC: 1:100, ICC/IF: 1:100

Type: Primary
Antigen: p23
Clonality: Monoclonal
Clone: JJ6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rabbit, Chicken, Guinea Pig, Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR