Anti-SHANK2 Mouse Monoclonal Antibody [clone: N23b/49]

Proveedor: ANTIBODIES.COM
A304967-100
ANTIA304967-100EA 475 EUR
ANTIA304967-100
Anti-SHANK2 Mouse Monoclonal Antibody [clone: N23b/49]
Anticuerpos
Mouse monoclonal [N23b/49] antibody to SHANK2 for WB, IHC, IP, ICC/IF and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, IP, ICC/IF, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: SHANK2
Clonality: Monoclonal
Clone: N23b/49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR