Anti-PFAS Rabbit Polyclonal Antibody

Proveedor: ANTIBODIES.COM
A307818-100
ANTIA307818-100EA 415 EUR
ANTIA307818-100
Anti-PFAS Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody to PFAS for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: PFAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR