Mouse Lamin B1 ELISA Kit

Proveedor: ANTIBODIES.COM

A310119-96
ANTIA310119-96EA 545 EUR
ANTIA310119-96
Mouse Lamin B1 ELISA Kit
Ensayos ELISAs

Mouse Lamin B1 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Lamin B1 in serum, plasma, and other biological fluids.


  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 7,5 to 120 ng/ml
  • Sensitivity: 0,26 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%


Mouse Lamin B1 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Lamin B1 in serum, plasma or other biological fluids. An antibody specific for Lamin B1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Lamin B1 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Lamin B1 antibody, which also binds the Lamin B1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Lamin B1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Lamin B1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Lamin B1 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Lamin B1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Información para pedidos: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

New Product

Order Now

ESPECIFICACIONES

Environmentally Preferable
Objetivo Lamin B1
Primary antibody reactivity Mouse
Tipo de ensayo Sandwich
Huésped
Formato Pre-coated
Duración del ensayo One step
Sensibilidad 0,26 ng/ml
Rango de detección 7,5 - 120 ng/ml
Método de detección Colorimetric
Tipo de la muestra Serum, plasma or other biological fluids
Volumen de la muestra 40 μl
Tiempo para resultados 1 h 30 min
Regulatory status RUO
Temperatura de almacenamiento 4 °C
Shelf life Store for 6 months at 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR