Human DHCR7 ELISA Kit

Proveedor: ANTIBODIES.COM

A313479-96
ANTIA313479-96EA 545 EUR
ANTIA313479-96
Human DHCR7 ELISA Kit
Ensayos ELISAs

Human DHCR7 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human DHCR7 in serum, plasma, and other biological fluids.


  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 150 to 2400 ng/L
  • Sensitivity: 9,28 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%


Human DHCR7 ELISA kit (A313479) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human DHCR7 in serum, plasma or other biological fluids. An antibody specific for DHCR7 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the DHCR7 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-DHCR7 antibody, which also binds the DHCR7 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-DHCR7 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-DHCR7 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of DHCR7 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of DHCR7 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Información para pedidos: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR