Human CYP2R1 ELISA Kit

Proveedor: ANTIBODIES.COM

A314311-96
ANTIA314311-96EA 545 EUR
ANTIA314311-96
Human CYP2R1 ELISA Kit
Ensayos ELISAs

Human CYP2R1 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CYP2R1 in serum, plasma, and other biological fluids.


  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 75 to 1200 ng/L
  • Sensitivity: 5,19 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%


Human CYP2R1 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human CYP2R1 in serum, plasma or other biological fluids. An antibody specific for CYP2R1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CYP2R1 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-CYP2R1 antibody, which also binds the CYP2R1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-CYP2R1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-CYP2R1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of CYP2R1 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of CYP2R1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Información para pedidos: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR