Human PSCA ELISA Kit

Proveedor: ANTIBODIES.COM

A78680-96
ANTIA78680-96EA 600 EUR
ANTIA78680-96
Human PSCA ELISA Kit
Ensayos ELISAs

Human PSCA ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PSCA in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready to use ELISA kit
  • Detection range: 0,313 to 20 ng/ml
  • Sensitivity: 0,188 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra assay: CV <8%, Inter assay: CV <10%


Human PSCA ELISA kit (A78680) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human PSCA in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PSCA has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PSCA present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PSCA Antibody, which binds the captured PSCA present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PSCA captured in each well. The concentration of PSCA can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Información para pedidos: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now

ESPECIFICACIONES

Objetivo PSCA
Environmentally Preferable
Primary antibody reactivity Human
Tipo de ensayo Sandwich
Principio de ensayo Quantitative
Huésped
Formato Pre-coated
Duración del ensayo Multiple steps
Sensibilidad 0,188 ng/ml
Rango de detección 0,313 - 20 ng/ml
Método de detección Colorimetric
Reactividad cruzada Human PSCA ELISA kit exhibits high specificity and excellent specificity for the detection of human PSCA. No significant cross-reactivity or interference between PSCA and analogues was observed.
Tipo de la muestra Serum, plasma, tissue homogenates and other biological fluids
Volumen de la muestra 100 μl
Tiempo para resultados 4 h 30 min
Regulatory status RUO
Temperatura de almacenamiento 4 °C
Shelf life Store for 6 months at 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR