Anti-SHP1 Rabbit Polyclonal Antibody

Proveedor: ANTIBODIES.COM
A94172-100
ANTIA94172-100EA 475 EUR
ANTIA94172-100
Anti-SHP1 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody to SHP-1 (phospho Tyr536) for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, IHC, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:5000

Type: Primary
Antigen: SHP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR