5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)

Proveedor: Apollo Scientific

BIC1074-100MG BIC1074-1G
APOSBIC1074-100MGEA 174 EUR
APOSBIC1074-100MG APOSBIC1074-1G
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)
Formula: C₂₅H₁₅NO₉
Número MDL: MFCD04119713
Núm. CAS: 117548-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR