N-Hydroxysulphosuccinimide

Proveedor: Apollo Scientific
Atención

Sinónimos: Sulpho NHS

BIM108-500MG BIM108-1G BIM108-5G
APOSBIM108-500MGEA 87 EUR
APOSBIM108-500MG APOSBIM108-1G APOSBIM108-5G
N-Hydroxysulphosuccinimide
N-Hydroxysulphosuccinimide

Forms amine reactive peptides


  • Protein


Ref: Staros,J.V,Wright,R.W.and Swingle,D.M.(1986) Anal.Biochem.156, 220-222

Formula: C₄H₅NO₆S
MW: 195.151 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00217065
Núm. CAS: 82436-78-0

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR