4-Methylumbelliferyl-2-acetamido-2-deoxy-6-sulphate-Β-D-glucopyranoside sodium salt

Proveedor: Apollo Scientific
Atención

BIM1189-25MG BIM1189-50MG
APOSBIM1189-25MGEA 255 EUR
APOSBIM1189-25MG APOSBIM1189-50MG
4-Methylumbelliferyl-2-acetamido-2-deoxy-6-sulphate-Β-D-glucopyranoside sodium salt
4-Methylumbelliferyl-2-acetamido-2-deoxy-6-sulphate-Β-D-glucopyranoside sodium salt

Fluorescent substrate for isoenzyme A of N-acetyl-b-D-glucosaminidase (Hexosaminidase A)

MW: 481.50 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00132972
Núm. CAS: 142439-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR