Acetyl pepstatin
Proveedor: Apollo Scientific
BIMI7332-25MG
APOSBIMI7332-25MGEA 40.6 EUR
APOSBIMI7332-25MG
Acetyl pepstatin
Proteínas y Péptidos
Inhibitor of aspartyl proteases, particularly effective onHIV-I proteinase and HIV-II proteinase