Tetraethylene glycol bis(p-toluenesulphonate)

Proveedor: Apollo Scientific

BIPG1869-10G BIPG1869-5G BIPG1869-25G
APOSBIPG1869-10GEA 157 EUR
APOSBIPG1869-10G APOSBIPG1869-5G APOSBIPG1869-25G
Tetraethylene glycol bis(p-toluenesulphonate)
Tetraethylene glycol bis(p-toluenesulphonate)
Formula: C₂₂H₃₀O₉S₂
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00075239
Núm. CAS: 37860-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR