AM-095 free acid 98%

Proveedor: Apollo Scientific

BISN0022-1MG BISN0022-5MG BISN0022-10MG
APOSBISN0022-1MGEA 109 EUR
APOSBISN0022-1MG APOSBISN0022-5MG APOSBISN0022-10MG
AM-095 free acid 98%
AM-095

AM095 free acid is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.

Formula: C₂₇H₂₄N₂O₅
Temperatura de almacenaje: Ambiente
Número MDL: MFCD22665734
Núm. CAS: 1228690-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR