L(+)-Diisopropyl tartrate

Proveedor: Apollo Scientific
Atención

Sinónimos: Diisopropyl (2R,3R)-2,3-dihydroxysuccinate, L-(+)-Tartaric acid diisopropyl ester

OR10051-100G OR10051-250G OR10051-500G
APOSOR10051-100GEA 42.2 EUR
APOSOR10051-100G APOSOR10051-250G APOSOR10051-500G
L(+)-Diisopropyl tartrate
L(+)-Diisopropyl tartrate

Asymmetric synthesis catalyst; Chiral intermediate for pharmaceuticals and agrochemicals.

Formula: C₁₀H₁₈O₆
MW: 234.25 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00064450
Núm. CAS: 2217-15-4

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR