Cyclobutyl methyl ketone

Proveedor: Apollo Scientific

OR304351-25G OR304351-5G
APOSOR304351-25GEA 533 EUR
APOSOR304351-25G APOSOR304351-5G
Cyclobutyl methyl ketone
Cyclobutyl methyl ketone
Formula: C₆H₁₀O
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00037148
Núm. CAS: 3019-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR