4H-Thieno[3,2-b]pyrrole-5-carboxylic acid hydrazide

Proveedor: Apollo Scientific

OR346266-1G
APOSOR346266-1GEA 265 EUR
APOSOR346266-1G
4H-Thieno[3,2-b]pyrrole-5-carboxylic acid hydrazide
4H-Thieno[3,2-b]pyrrole-5-carboxylic acid hydrazide
Formula: C₇H₇N₃OS
MW: 181,22 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD04208027
Núm. CAS: 119448-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR