2-Morpholineacetic acid hydrochloride 97%

Proveedor: Apollo Scientific

OR452078-1G OR452078-250MG OR452078-500MG
APOSOR452078-1GEA 654 EUR
APOSOR452078-1G APOSOR452078-250MG APOSOR452078-500MG
2-Morpholineacetic acid hydrochloride 97%
2-Morpholineacetic acid hydrochloride
Formula: C₆H₁₁NO₃·ClH
Temperatura de almacenaje: Ambiente
Número MDL: MFCD07779523
Núm. CAS: 1187929-25-4

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR