[(5-Methyl-1,3-thiazol-2-yl)methyl]amine dihydrochloride

Proveedor: Apollo Scientific

OR470664-1G OR470664-250MG
APOSOR470664-1GEA 632 EUR
APOSOR470664-1G APOSOR470664-250MG
[(5-Methyl-1,3-thiazol-2-yl)methyl]amine dihydrochloride
[(5-Methyl-1,3-thiazol-2-yl)methyl]amine dihydrochloride
Formula: C₅H₁₀Cl₂N₂S
Temperatura de almacenaje: Nevera
Número MDL: MFCD18071410
Núm. CAS: 921091-08-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR