Methyl 2-Acetamido-3-(3-pyridyl)propanoate

Proveedor: Apollo Scientific

OR470693-1G
APOSOR470693-1GEA 1120 EUR
APOSOR470693-1G
Methyl 2-Acetamido-3-(3-pyridyl)propanoate
Methyl 2-Acetamido-3-(3-pyridyl)propanoate
Formula: C₁₁H₁₄N₂O₃
MW: 222,24 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD07700262
Núm. CAS: 1064157-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR