Methyl 2-bromo-3-methyl-5-nitrobenzoate ≥95%

Proveedor: Apollo Scientific

OR60272-250MG OR60272-1G OR60272-5G
APOSOR60272-250MGEA 87 EUR
APOSOR60272-250MG APOSOR60272-1G APOSOR60272-5G
Methyl 2-bromo-3-methyl-5-nitrobenzoate ≥95%
Methyl 2-bromo-3-methyl-5-nitrobenzoate
Formula: C₉H₈BrNO₄
Número MDL: MFCD22381695
Núm. CAS: 179897-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR