5-Bromo-4-chloro-3-indolyl-α-D-N-acetylneuraminic acid
Proveedor: Apollo Scientific
Atención
OR8230T-5MG
APOSOR8230T-5MGEA 176 EUR
APOSOR8230T-5MG
5-Bromo-4-chloro-3-indolyl-α-D-N-acetylneuraminic acid
5-Bromo-4-chloro-3-indolyl-α-D-N-acetylneuraminic acid
A substrate for the detection of sialidase-like enzyme,screening of enzymes, studying the physiological role of gangliosides, & for recombinant technologies.