Methyl 4,5,6,7-tetrahydro-1H-benzimidazole-5-carboxylate 96%

Proveedor: Apollo Scientific

OR910353-1G
APOSOR910353-1GEA 502 EUR
APOSOR910353-1G
Methyl 4,5,6,7-tetrahydro-1H-benzimidazole-5-carboxylate 96%
Methyl 4,5,6,7-tetrahydro-1H-benzimidazole-5-carboxylate
Formula: C₉H₁₂N₂O₂
MW: 180,204 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD20662345
Núm. CAS: 1437794-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR