(R)-1-Chloropropan-2-ol 98%

Proveedor: Apollo Scientific
Peligro

OR911848-250MG OR911848-1G OR911848-5G
APOSOR911848-250MGEA 58 EUR
APOSOR911848-250MG APOSOR911848-1G APOSOR911848-5G
(R)-1-Chloropropan-2-ol 98%
(R)-1-Chloropropan-2-ol
Formula: C₃H₇ClO
Número MDL: MFCD01862132
Núm. CAS: 19141-39-0
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR