trans-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate 95%

Proveedor: Apollo Scientific

OR917667-1G OR917667-250MG
APOSOR917667-1GEA 89.9 EUR
APOSOR917667-1G APOSOR917667-250MG
trans-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate 95%
trans-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate
Formula: C₉H₁₆O₃
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03840884
Núm. CAS: 110928-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR