Chloromethyl cyclohexyl ether 95%

Proveedor: Apollo Scientific

Sinónimos: (Chloromethoxy)cyclohexane

OR921970-25G
APOSOR921970-25GEA 184 EUR
APOSOR921970-25G
Chloromethyl cyclohexyl ether 95%
Chloromethyl cyclohexyl ether
Formula: C₇H₁₃ClO
MW: 148.63 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD10566884
Núm. CAS: 3587-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR