Ethynyl-p-tolyl sulphone 95%

Proveedor: Apollo Scientific

Sinónimos: Tosylacetylene, Ethynyl p-Tolyl sulphone, p-Toluenesulphonylacetylene

OR923266-1G
APOSOR923266-1GEA 95.7 EUR
APOSOR923266-1G
Ethynyl-p-tolyl sulphone 95%
Ethynyl-p-tolyl sulphone
Formula: C₉H₈O₂S
MW: 180.22 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00191647
Núm. CAS: 13894-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR