trans-4-Metilciclohexanol 97%

Proveedor: Apollo Scientific

OR924640-5G
APOSOR924640-5GEA 136 EUR
APOSOR924640-5G
trans-4-Metilciclohexanol 97%
trans-4-Metilciclohexanol
Formula: C₇H₁₄O
Número MDL: MFCD00064171
Núm. CAS: 7731-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR