Benzoyl-β-alanine 98%

Proveedor: Apollo Scientific

OR924812-25G OR924812-100G
APOSOR924812-25GEA 45.1 EUR
APOSOR924812-25G APOSOR924812-100G
Benzoyl-β-alanine 98%
Benzoyl-beta-alanine
Formula: C₁₀H₁₁NO₃
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00037293
Núm. CAS: 3440-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR