N-Succinimidyl Methacrylate 95%

Proveedor: Apollo Scientific

Sinónimos: N-Methacryloyloxysuccinimide, Methacrylic Acid N-Succinimidyl Ester

OR937503-5G
APOSOR937503-5GEA 142 EUR
APOSOR937503-5G
N-Succinimidyl Methacrylate 95%
N-Succinimidyl Methacrylate
Formula: C₈H₉NO₄
MW: 183.16 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD15072193
Núm. CAS: 38862-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR