O-Acetyl-N-tert-butylhydroxylamine hydrochloride 95%

Proveedor: Apollo Scientific

Sinónimos: O-Acetyl-N-(tert-butyl)hydroxylamine hydrochloride

OR938608-1G
APOSOR938608-1GEA 128 EUR
APOSOR938608-1G
O-Acetyl-N-tert-butylhydroxylamine hydrochloride 95%
O-Acetyl-N-tert-butylhydroxylamine hydrochloride
Formula: C₆H₁₃NO₂
MW: 167,63 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD12545957
Núm. CAS: 851074-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR