(2E,4E)-5-Phenyl-penta-2,4-dienoic acid methyl ester 95%

Proveedor: Apollo Scientific

OR955546-1G
APOSOR955546-1GEA 632 EUR
APOSOR955546-1G
(2E,4E)-5-Phenyl-penta-2,4-dienoic acid methyl ester 95%
(2E,4E)-5-Phenyl-penta-2,4-dienoic acid methyl ester
Formula: C₁₂H₁₂O₂
MW: 188,222 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09033558
Núm. CAS: 1516-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR