H-D-Lys(Z)-OH 97%

Proveedor: Apollo Scientific
Atención

Sinónimos: H-D-Lys(Cbz)-OH, Nepsilon-Benzyloxycarbonyl-D-lysine, Nepsilon-Cbz-D-lysine, H-D-Lys(Z)-OH, Nepsilon-Carbobenzoxy-D-lysine

OR964205-100G OR964205-25G
APOSOR964205-100GEA 400 EUR
APOSOR964205-100G APOSOR964205-25G
H-D-Lys(Z)-OH 97%
H-D-Lys(Z)-OH
Formula: C₁₄H₂₀N₂O₄
MW: 280.324 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00063120
Núm. CAS: 34404-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR