Methyl 3-pyridylacetate hydrochloride 95%

Proveedor: Apollo Scientific

OR965088-1G OR965088-5G
APOSOR965088-1GEA 168 EUR
APOSOR965088-1G APOSOR965088-5G
Methyl 3-pyridylacetate hydrochloride 95%
Methyl 3-pyridylacetate hydrochloride
Formula: C₈H₉NO₂·ClH
Temperatura de almacenaje: Ambiente
Número MDL: MFCD19442777
Núm. CAS: 69966-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR