Potassium cyclohexylmethyl trifluoroborate

Proveedor: Apollo Scientific
Peligro

PC430518-500MG PC430518-1G PC430518-5G
APOSPC430518-500MGEA 0 EUR
APOSPC430518-500MG APOSPC430518-1G APOSPC430518-5G
Potassium cyclohexylmethyl trifluoroborate
Potassium cyclohexylmethyl trifluoroborate
Formula: C₇H₁₃BF₃·K
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11052658
Núm. CAS: 1346647-18-4
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR