Benzoic acid N'-(6-chloro-4-trifluoromethyl-pyridin-2-yl)-N'-methyl-hydrazide

Proveedor: Apollo Scientific

PC446077-100MG
APOSPC446077-100MGEA 265 EUR
APOSPC446077-100MG
Benzoic acid N'-(6-chloro-4-trifluoromethyl-pyridin-2-yl)-N'-methyl-hydrazide
Benzoic acid N'-(6-chloro-4-trifluoromethyl-pyridin-2-yl)-N'-methyl-hydrazide
Formula: C₁₄H₁₁ClF₃N₃O
MW: 329.705 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD24386176
Núm. CAS: 1431555-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR