Silver bis(trifluoromethanesulphonyl)imide

Proveedor: Apollo Scientific

PC50957-1G PC50957-5G PC50957-25G
APOSPC50957-1GEA 0 EUR
APOSPC50957-1G APOSPC50957-5G APOSPC50957-25G
Silver bis(trifluoromethanesulphonyl)imide
Silver bis(trifluoromethanesulphonyl)imide
Formula: C₂AgF₆NO₄S₂
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08457644
Núm. CAS: 189114-61-2
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR