(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) )

Proveedor: Apollo Scientific

Sinónimos: ((3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), 7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, (7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium Hexafluorophosphate, (7-Azabenzotriazol-1-yl)-N-oxy-tris(dimethyamino)phosphonium hexafluorosphate, AOP, Tris(dimethylamino)(3H-1,2,3-triazolo[4,5-b]pyridin-3-yloxy)phosphonium Hexafluorophosphate

PC510112-25G
APOSPC510112-25GEA 60.9 EUR
APOSPC510112-25G
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) )
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) )
Formula: C₁₂H₂₂F₆N₆OP₂
MW: 443.27 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09263289
Núm. CAS: 156311-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR