2-Bromo-6-methylphenyl trifluoromethanesulphonate 95%

Proveedor: Apollo Scientific
Peligro

PC51196-1G PC51196-250MG PC51196-5G
APOSPC51196-1GEA 140 EUR
APOSPC51196-1G APOSPC51196-250MG APOSPC51196-5G
2-Bromo-6-methylphenyl trifluoromethanesulphonate 95%
2-Bromo-6-methylphenyl trifluoromethanesulphonate
Formula: C₈H₆BrF₃O₃S
Temperatura de almacenaje: Ambiente
Número MDL: MFCD27975433
Núm. CAS: 1820716-91-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR