H1 oat medium
Proveedor: Apollo Scientific
PMO612B-10L
APOSPMO612B-10LEA 52.2 EUR
APOSPMO612B-10L
H1 oat medium
Medios de cultivo celular
Contains agar and sucrose. Contains the macro- and micronutrients and supplements required for the symbiotic culture of orchids.
  • Plant tissue culture tested