Culture media, in Petri dishes, Slanetz Bartley medium
Proveedor: PanReac AppliChem
443812.0922
APPC443812.0922EA 99.9 EUR
APPC443812.0922
Culture media, in Petri dishes, Slanetz Bartley medium
Medios Medios preparados
Prepared medium for the enumeration of Enterococci.
  • Pinkish beige