Anti-COPA Rabbit Polyclonal Antibody

Proveedor: Aviva Systems Biology
ARP51894_P050
AVIVARP51894_P050EA 401 EUR
AVIVARP51894_P050
Anti-COPA Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: COPA (coatomer protein complex, subunit alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR