Anti-ERCC6L2 Rabbit Polyclonal Antibody

Proveedor: Aviva Systems Biology
ARP54402_P050
AVIVARP54402_P050EA 401 EUR
AVIVARP54402_P050
Anti-ERCC6L2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ERCC6L2 (excision repair cross-complementation group 6-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

ESPECIFICACIONES

Simbolo antígeno ERCC6L2
Nombre del antígeno Excision repair cross-complementation group 6-like 2
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad
Huésped Rabbit
Isotipo
Western blot Yes
Epítopo C-terminal
Adsorción cruzada none
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR