Anti-SLX4 Rabbit Polyclonal Antibody

Proveedor: Aviva Systems Biology
ARP59857_P050
AVIVARP59857_P050EA 401 EUR
AVIVARP59857_P050
Anti-SLX4 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SLX4 (SLX4 structure-specific endonuclease subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno SLX4
Environmentally Preferable
Nombre del antígeno SLX4 structure-specific endonuclease subunit
Sinónimos de antígenos MUS312|FANCP|BTBD12
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human
Huésped Rabbit
ID del gen 84464
Isotipo
Western blot Yes
Epítopo N-terminal
Adsorción cruzada none
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR