Pre-sterilized individually wrapped blue pestle, Axygen®
Proveedor: Corning
PES-15-B-SI
AXYGPES-15-B-SIEA 569 EUR
AXYGPES-15-B-SI
Pre-sterilized individually wrapped blue pestle, Axygen®
Morteros y manos de mortero Manos de mortero
Durable and inert polypropylene construction
  • Designed to fit 1.5 and 2.0 ml centrifuge tubes
  • Individually wrapped and sterilized